Coating thickness gauge, PosiPen
Fournisseur: Industrial Physics
LD6000
SIMELD6000EA 330 EUR InStock
SIMELD6000
Coating thickness gauge, PosiPen
Contrôle des revêtements
This thickness gauge is designed for the measurement of non magnetic coatings include paint, enamel, plating, hot-dip galvanising on steel. A small magnet can be placed with pin-point accuracy on small parts and on peaks and valleys.
  • Not affected by mechanical shock, acid, oil, water and dust making it rugged
  • Highly wear resistant probe tip for long life and continuous accuracy
  • Two scales: Mils (inch) and microns (metric)
Used for small, hot or hard-to-reach surfaces, spot checks and sprayed coatings. For use in refineries, petrochemical plants, electroplating, chrome, zinc, galvanizing platers and powder coaters.
Information de livraison: Supplied with a leather pouch, a certificate of calibration (traceable to NIST) and instructions.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité