Thickness gauges
Fournisseur: Industrial Physics
LD7016
SIMELD7016EA 602 EUR InStock
SIMELD7016
Thickness gauges
Contrôle des revêtements
These thickness gauges are designed to measure the thickness of metallic and non-metallic materials (aluminium, titanium, plastics, ceramics, glass, plastics). It can monitor pipes and vessels for loss of thickness due to corrosion or erosion.
  • Large screen, battery indicator and a coupling indicator
  • Automatic power shut down, simple calibration procedure
  • Not suitable for cast iron due to its big crystalloid composition
'Basic' model has ten preset sound velocities.
'Pro' version allows for the storage of data (10 storage units) and performs a sound velocity measurement thanks to a known material thickness.
Information de livraison: Supplied with ultrasonic gel (60 ml), probe (10 mm, 5,0 MHz), calibration block and manual. 'Pro' version has an extra probe (10 mm, 2,5 MHz).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité