UV inspection lamp
Fournisseur: Industrial Physics
LD7220
SIMELD7220EA 675 EUR InStock
SIMELD7220
UV inspection lamp
Éclairages Lampes UV
This robust, handheld rechargeable inspection spotlight is used to detect contaminations that react under UV-illumination and cannot be seen with naked eye such as some organic fats and alkaline contaminants. Ideal to inspect the cleanliness of steel prior to painting.
  • Equipped with high power UV LED (450 mW at 365 nm wavelength)
  • Anodised aluminium lamp body is resistant to corrosion and stands up to years of heavy use
  • Single wavelength, no heat but deep UV light
  • Re-chargeable Li-ion battery with 180 min. life time
Output UV light intensity >1500 µW/cm² at 38 cm
Runtime (fully charged): approximately 3 hours
LED lifetime: 20000 hours
Weight: 190 g (without battery)
Information de livraison: Supplied in a sturdy plastic suitcase complete with yellow safety glasses for optimal contrast, Li-ion battery, battery charger and carrying case.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité