SP7310
SIMESP7310EA
584
EUR
InStock
SIMESP7310
Test kit, chloride test, Bresle method
Kits de tests environnementaux
TQC Bresle kit is a chloride test kit that complies with the ISO 8502-6 and ISO 8502-9 standards. The Bresle method is designed to to assess the level of soluble salts using a Bresle patch or Bresle sampler, distilled water and a conductivity meter. The conductivity is mainly directly proportional to the concentration of dissolved chloride ions in the solution. The kit includes all the necessary equipment to execute a Bresle test that will indicate the contamination of soluble salts on blast-cleaned surfaces prior to coating. Included is a conductivity gauge used for the assessment of soluble salt ions as chlorides, sulphates and nitrates.
- TQC DSP-procedure eliminates user-errors and saves time
- Compact and complete kit in sturdy case
- Unique Direct Sampling Procedure (DSP) to ensure high speed and accuracy
- Up to 60 times more accurate than other test kits available
Contamination of blast media
It is also suitable to determine the contamination of blast-media in use. This important test described in the ISO 11127-6 and ISO 11127-7 standards helps to prevent the dissolved salts in the recycled abrasive media or water re-contaminating the surface being cleaned.
Normalisation temperature: 25 °C
Information de livraison: Kit supplied with digital conductivity meter, 25×Bresle patch, distilled water, cups and syringes, calibration and cleansing solutions, manual ( pictorial) and case.