Strips of 8 tubes
Fournisseur: Simport Scientific
T110-15
SIMPT110-15EA 25.5 EUR InStock
SIMPT110-15
Strips of 8 tubes
Tubes Microtubes
Made of PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Certified RNase, DNase, Pyrogen and DNA-free
These plates feature 600 µl wells for smaller volume applications. They include a deep well plate along with twelve eight-tube strips. The tube strips are removable and can also be ordered separately.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité