Sterilin™ Scintillation Vials with Cap
Fournisseur: STERILIN
S31
SLINS31EA 596 EUR InStock
SLINS31
Sterilin™ Scintillation Vials with Cap
Fioles
Sterilin® Scintillation vials are ideal for use in liquid scintillation counting, beta/gamma counting and for sample storage. Vials are made of PP and caps are PE. The quarter-turn cap ensures reliable sample storage.
  • Precision manufactured for strength and chemical resistance
  • Quarter turn screw cap ensures excellent sample containment
  • 6 ml vial with flush fitting cap is ideal as insert for scintillation vials
  • Not available in all geographic regions
Certifications: Certificate of Analysis (COA), Certificate of Conformance (COC).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité