Laboratory Glassware Washers, GW4290
Fournisseur: SMEG
GW4290-00-000 GW4290-S0-000 GW4290-SC-000
SMEGGW4290-00-000EA 0 EUR InStock
SMEGGW4290-00-000 SMEGGW4290-S0-000 SMEGGW4290-SC-000
Laboratory Glassware Washers, GW4290
Lave-verres
The GW4290 90 cm glass washer is equipped with a soft touch colour control panel that allows simple and intuitive daily use.
  • Colour touch display
  • Detergent compartment
  • Maximum loading flexibility
  • Data recording
  • Complete drying
The compact design of the interface, which gives linearity to the surface, does not neglect the completeness of the information: the operator has at his disposal a three-level range of settings complete with 20 pre-set programmes and 20 fully customisable ones to satisfy every need.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité