Antibody labelling kits, Antibody-Oligonucleotide All-in-One conjugation kit
Fournisseur: TRILINK BIOTECHNOLOGIES
A-9202-001
SOLUA-9202-001EA 1096 EUR InStock
SOLUA-9202-001
Antibody labelling kits, Antibody-Oligonucleotide All-in-One conjugation kit
Tests Essais protéiniques
The Antibody-Oligonucleotide All-in-One Conjugation kit offers TurboLink™ catalysed conjugation based on Solulink’s proprietary linking technology to prepare antibody-oligonucleotide conjugates without chromatography. The kit includes buffers, spin columns, and a calculator to determine MSR. No column chromatography is required. With the Antibody-Oligonucleotide All-in-One Conjugation kit, you can generate high-purity conjugates virtually free of residual antibody or oligonucleotide (>95% conjugate).
  • Simple and easy to use – Requires only micropipette, microcentrifuge, and UV spectrophotometer
  • Automated calculations – Calculator with fully integrated input/output provided
  • High Yield – 30 to 50% yield based on starting antibody
  • High Purity – >95% purity without chromatographic purification
  • High stability – Conjugates are stable for >1 year
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité