Biotinylation kits, ChromaLink™ One-Shot antibody labelling kit
Fournisseur: TRILINK BIOTECHNOLOGIES
B-9007-020K
SOLUB-9007-020KEA 0 EUR InStock
SOLUB-9007-020K
Biotinylation kits, ChromaLink™ One-Shot antibody labelling kit
Tests Essais protéiniques
ChromaLink™ Biotin contains a UV-traceable chromophore to enable reproducibility labelling. The degree of biotinylation can be measured in minutes, without the standard curves required for HABA/avidin and fluoro-reporter assays. With a simple and direct UV scan, biotin incorporation can be quantified and reproducible production of consistent batches ensured. ChromaLink™ labelling can be paired with NanoLink™ Streptavidin Magnetic Beads for many types of assay development.
  • Reproducible production – UV-traceable chromophore permits non destructive, rapid A280/A354 quantification of incorporated biotin, enabling consistency and reproducibility
  • Extended PEG3 spacer – helps reduce aggregation, minimises steric hindrance, and enhances solubility
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité