Antibody labelling kits, Fluorescein one-shot antibody labelling kit
Fournisseur: TRILINK BIOTECHNOLOGIES
F-9001-009K
SOLUF-9001-009KEA 0 EUR InStock
SOLUF-9001-009K
Antibody labelling kits, Fluorescein one-shot antibody labelling kit
Tests Essais protéiniques
Fluorescein Antibody Conjugation kit offers an efficient method that delivers >95% conversion of antibody to conjugate. The kit includes everything needed to make two direct-labelled fluorescein-antibody conjugates from 100 µg of any user-supplied antibody.
  • Fast – Method generates conjugates in just 90 min
  • High purity – Easy spin-column purification columns deliver highly purified conjugates
  • Gentle – Amine-reactive chemistry leaves antibodies intact - no reducing reagents
  • Easy to use – Protocols and calculators provided to determine MSR
  • Versatile – Compatible with Western, ELISA, or IHC applications
  • Efficient – 100% conversion of antibody to conjugate with high (>70%) purification yields
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité