Antibody labelling kits, R-PE-Antibody Conjugation kit
Fournisseur: TRILINK BIOTECHNOLOGIES
P-9002-002
SOLUP-9002-002EA 966 EUR InStock
SOLUP-9002-002
Antibody labelling kits, R-PE-Antibody Conjugation kit
Tests Essais protéiniques
The R-PE-Antibody Conjugation kit offers an innovative catalysed conjugation that delivers >95% conversion of antibody to conjugate. The kit includes everything needed to make two direct-labelled R-PE antibody conjugates from 500 µg of any user-supplied antibody.
  • Catalysed conjugation – Fast catalysed method generates conjugates quickly and efficiently
  • Quantifiable – Conjugate bond is UV-traceable for simple and direct quantification
  • High purity – Easy spin-column purification columns deliver highly purified conjugates
  • Gentle – Amine-reactive chemistry leaves antibodies intact - no reducing reagents
  • Easy to use – Protocols and calculators provided to determine MSR
  • Versatile – Compatible with western, ELISA, or IHC applications
  • Efficient – 95 to 100% conversion with high purification yields
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité