MA164-500GM
SPCMMA164-500GMEA
183
EUR
InStock
SPCMMA164-500GM
Manganèse (II) sulfate monohydraté ≥98.0%, en poudre BP, FCC
Manganèse (II) sulfate monohydraté
Manganese Sulfate, Monohydrate, Powder, FCC, BP - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Health hazard.