Nitrile NPT gloves, Vigor®
Fournisseur: SRI TRANG GLOVES
7028581
SRIT7028581EA 33 EUR InStock
SRIT7028581
Nitrile NPT gloves, Vigor®
Gants
High quality, dual-labelled PPE and MDR nitrile gloves.
  • Non sterile, powder-free, textured finish
  • Highly resistant to chemical solvents and fats
  • Excellent grip, especially when holding wet objects
  • Particularly skin friendly for users with latex allergy
  • Rolled cuffs minimise the risk of tearing
  • Level 2 – AQL 1,5
These durable, ambidextrous gloves are ideal for use in pharmaceutical and industrial lab applications, as well as applications in medical, manufacturing and cleaning areas. They are free from phthalates, softeners and allergenic latex proteins.
Certifications: EN 420, EN 455, EN ISO 374-1, EN 374-2, EN 16523-1, EN 374-4, EN 374-5; PPE Cat. III, Type B (KTP).
Food contact approved.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité