ALDEFLUOR™ DEAB reagent 1.5 mM , ALDH enzyme inhibitor
Fournisseur: STEMCELL Technologies
01705
STMC01705EA 164 EUR InStock
STMC01705
ALDEFLUOR™ DEAB reagent 1.5 mM , ALDH enzyme inhibitor
ALDEFLUOR™ DEAB reagent
ALDH enzyme inhibitor used as a control in the ALDEFLUOR™ assay system.
Use ALDEFLUOR™ DEAB Reagent as a negative control to account for background fluorescence in the ALDEFLUOR™ assay system.
Available either individually or as part of the ALDEFLUOR™ Kit, this diethylaminobenzaldehyde (DEAB) reagent is an aldehyde dehydrogenase (ALDH) enzyme inhibitor and can be used during the identification of ALDH-bright (ALDHbr) cells.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité