L'expérience en ligne d'Avantor évolue pour mieux vous accompagner ! À compter du 4 avril 2025, nos clients seront migrés vers une nouvelle plateforme pour une expérience d'achat en ligne simplifiée.
Agar Leukocyte Conditioned Medium
Fournisseur: STEMCELL Technologies
02300
STMC02300EA 596 EUR InStock
STMC02300
Agar Leukocyte Conditioned Medium
Milieux de culture cellulaire
Source of colony-stimulating factors for assays of human hematopoietic progenitor cells.
Agar Leukocyte Conditioned Medium (Agar-LCM) is a source of colony-stimulating factors for assays of hematopoietic progenitor cells from light-density fractions of human bone marrow and peripheral blood. It is suitable for the growth of colony-forming unit erythroid (CFU­E), burst-forming unit-erythroid (BFU-E), colony-forming unit-macrophage (CFU-M), colony-forming unit-granulocyte (CFU-G), colony-forming unit-granulocyte, macrophage (CFU-GM) and colony-forming unit-granulocyte, erythroid, macrophage, megakaryocyte (CFU-GEMM) when added to MethoCult™ medium at a final concentration of 10% with the addition of erythropoietin. Agar-LCM is prepared using normal human peripheral blood leukocytes in an agar-containing medium.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité