L'expérience en ligne d'Avantor évolue pour mieux vous accompagner ! À compter du 4 avril 2025, nos clients seront migrés vers une nouvelle plateforme pour une expérience d'achat en ligne simplifiée.
StemSpan™ Myeloid Expansion Supplement (100X)
Fournisseur: STEMCELL Technologies
02693
STMC02693EA 759 EUR InStock
STMC02693
StemSpan™ Myeloid Expansion Supplement (100X)
Milieux de culture cellulaire
Serum-free culture supplement for expansion and differentiation of human granulocytes.
StemSpan™ Myeloid Expansion Supplement (100X) contains a combination of recombinant human cytokines formulated to selectively promote the expansion and differentiation of human myeloid progenitor cells, generating large numbers of CD15+ granulocytes from CD34+ cells isolated from human cord blood (CB) or bone marrow (BM) samples.
StemSpan™ Myeloid Expansion Supplement is intended for use in combination with any of the following StemSpan™ media:
StemSpan™ SFEM
StemSpan™ SFEM II
StemSpan™-XF
StemSpan™-AOF
When added to serum-free medium, StemSpan™ Myeloid Expansion Supplement typically promotes the production of thousands of CD15+ granulocytes per input CD34+ cell in 14-day liquid cultures initiated with CD34+ human CB cells. See data tab for more details.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité