MethoCult™ M3134 Medium
Fournisseur: STEMCELL Technologies
03134
STMC03134EA 112 EUR InStock
STMC03134
MethoCult™ M3134 Medium
Milieux de culture cellulaire
Base methylcellulose medium for mouse cells.
MethoCult™ M3134 is an incomplete medium that contains 2.6% methylcellulose in Iscove's MDM. MethoCult™ M3134 is suitable for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of mouse bone marrow, spleen, peripheral blood, and fetal liver, when the appropriate growth factors and supplements are added. This formulation does not contain serum or cytokines.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité