MethoCult™ M3630 Medium
Fournisseur: STEMCELL Technologies
03630
STMC03630EA 533 EUR InStock
STMC03630
MethoCult™ M3630 Medium
Milieux de culture cellulaire
Methylcellulose-based medium for mouse pre-B lymphoid progenitor cells.
MethoCult™ M3630 is optimised for the growth and enumeration of mouse pre-B lymphoid progenitor cells in colony-forming unit (CFU) assays of mouse bone marrow and Whitlock-Witte long-term cultures.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité