MethoCult™ GF R3774 Medium
Fournisseur: STEMCELL Technologies
03774
STMC03774EA 780 EUR InStock
STMC03774
MethoCult™ GF R3774 Medium
Milieux de culture cellulaire
Methylcellulose-based medium with recombinant cytokines (without EPO) for rat cells.
MethoCult™ GF R3774 is optimised for growth and enumeration of granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M) in colony-forming unit (CFU) assays of rat bone marrow cells.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité