ClonaCell™-HY PEG Reagent
Fournisseur: STEMCELL Technologies
03806
STMC03806EA 91 EUR InStock
STMC03806
ClonaCell™-HY PEG Reagent
Milieux de culture cellulaire
Polyethylene glycol reagent for hybridoma fusion.
ClonaCell™-HY PEG is suitable for inducing cellular fusion of myeloma cells with lymphocytes to produce hybridomas. This reagent has been verified for use during mouse and rat hybridoma development and reportedly is compatible for production, cloning, and expansion of hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster. ClonaCell™-HY PEG is supplied as a 50% (w/v) solution of PEG 3350 in Dulbecco's Modified Eagle's Medium (DMEM) with gentamicin and phenol red. The solution has been pre-tested and qualified for cell fusion.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité