ClonaCell™-CHO CD Liquid Medium
Fournisseur: STEMCELL Technologies
03817
STMC03817EA 176 EUR InStock
STMC03817
ClonaCell™-CHO CD Liquid Medium
Milieux de culture cellulaire
Chemically defined, animal component-free, serum-free, protein-free, glutamine-free liquid medium for culture of CHO cells.
ClonaCell™-CHO CD Liquid Medium is a general purpose liquid medium for culture of suspension-adapted CHO cells. This chemically defined liquid medium is protein-free and animal component-free. It does not contain L-glutamine, selection agents, antibiotics, or phenol red. ClonaCell™-CHO CD Liquid Medium is compatible with a variety of selection systems, including dihydrofolate reductase (DHFR) and glutamine synthetase (GS).
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité