ClonaCell™ Expansion Medium
Fournisseur: STEMCELL Technologies
03835
STMC03835EA 411 EUR InStock
STMC03835
ClonaCell™ Expansion Medium
Milieux de culture cellulaire
Hybridoma expansion medium with hypoxanthine and thymidine (animal origin-free).
ClonaCell™-HY AOF Expansion Medium is an animal origin-free (AOF) and serum-free liquid medium optimised for hybridoma expansion after hypoxanthine, aminopterin, thymidine (HAT) selection. The medium contains hypoxanthine and thymidine (HT) and is used to wean hybridomas off aminopterin used during the selection process. ClonaCell™-HY AOF Expansion Medium promotes robust expansion of hybridomas, adaptation from serum-containing to serum-free media, and stabilising the viability and antibody productivity of hybridoma cell lines. The majority of hybridomas can be switched directly from serum-containing media to ClonaCell™-HY AOF Expansion Medium without an adaptation step.
The absence of serum in this medium facilitates detection and purification of the desired hybridoma-derived antibody without interference from serum-derived immunoglobulins. This medium has been verified for use with mouse and rat hybridomas and is suitable for expansion of mouse myelomas such as Sp2/0, rat myelomas such as YB2/0, and human myelomas such as U266. In most cases, cryopreserved hybridomas can be thawed directly into ClonaCell™-HY AOF Expansion Medium while maintaining a high level of viability. This medium may be used as a serum-free alternative to ClonaCell™-HY Medium A or ClonaCell™-HY Medium E. No materials of animal or human origin are used in the manufacture of this medium or its components, to at least the secondary level of manufacturing.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité