MethoCult™ H4035 Optimum without EPO
Fournisseur: STEMCELL Technologies
04035
STMC04035EA 590 EUR InStock
STMC04035
MethoCult™ H4035 Optimum without EPO
Milieux de culture cellulaire
Methylcellulose-based medium with recombinant cytokines (without EPO) for human cells. 1×100 ml bottle.
MethoCult™ H4035 Optimum Without EPO (MethoCult™ GF H4035) is a methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples. MethoCult™ H4035 Optimum Without is formulated to support optimal growth of granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M). The inclusion of G-CSF in Optimum formulations improves discrimination of myeloid colony subtypes. This formulation is compatible with STEMvision™ software for automated colony counting.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité