MethoCult™ SF H4636 Medium
Fournisseur: STEMCELL Technologies
04636
STMC04636EA 603 EUR InStock
STMC04636
MethoCult™ SF H4636 Medium
Milieux de culture cellulaire
Serum-free methylcellulose-based medium with recombinant cytokines for human ES and iPS cell-derived cells.
MethoCult™ SF H4636 is recommended for the culture of human embryonic stem (ES) cell-derived and induced pluripotent stem (iPS) cell-derived hematopoietic progenitor cells in defined serum-free conditions. MethoCult™ SF H4636 is formulated to support optimal growth of erythroid progenitor cells (CFU-E and BFU-E), granulocyte-macrophage progenitor cells (CFU-GM, CFU-G, CFU-M), and multipotential progenitor cells (CFU-GEMM; granulocyte, erythrocyte, macrophage, megakaryocyte). MethoCult™ SF H4636 is also recommended for CFU assays with mononuclear cells, CD34+ enriched cells, and cells isolated by other purification methods from human bone marrow (BM), mobilised peripheral blood (MPB), peripheral blood (PB), and cord blood (CB) samples.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité