MegaCult™ C Medium with Lipids
Fournisseur: STEMCELL Technologies
04850
STMC04850EA 897 EUR InStock
STMC04850
MegaCult™ C Medium with Lipids
Milieux de culture cellulaire
Medium with lipids, without cytokines.
MegaCult™-C Medium with Lipids is intended for the culture of CFU-Mk in human bone marrow, mobilized peripheral blood, and cord blood samples after addition of appropriate cytokines. It is recommended for use with CD34+-enriched cells, mononuclear cells, and cells isolated by other purification methods. It can also be used for CFU-MK assays of megakaryocyte progenitor cells in unseparated or purified cell suspensions of mouse bone marrow after addition of appropriate cytokines.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité