Collagen Solution
Fournisseur: STEMCELL Technologies
04902
STMC04902EA 1422 EUR InStock
STMC04902
Collagen Solution
Suppléments et additifs pour culture cellulaire
For preparation of collagen gels and for coating cell culture surfaces - 35 ml.
Enhance cell attachment, growth and proliferation with collagen solution. This natural extracellular matrix protein provides structural support and mimics physiological conditions to promote cell adhesion, expansion, migration, and differentiation. Use this versatile cell culture surface to prepare collagen gels and to coat plasticware for routine cell culture.
Collagen solution is compatible for use with MegaCult™-C for the culture of human or mouse megakaryocytic progenitor cells, and can also be purchased as a kit with MegaCult™-C medium with cytokines (STMC04961).
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité