MegaCult™-C Collagen and Medium with Cytokines
Fournisseur: STEMCELL Technologies
04961
STMC04961EA 2957 EUR InStock
STMC04961
MegaCult™-C Collagen and Medium with Cytokines
Milieux de culture cellulaire
Kit containing collagen and medium with cytokines for human CFU-Mk assays.
MegaCult™-C collagen and medium with cytokines kit includes includes medium and collagen solution necessary for performing colony-forming unit (CFU) assays of human megakaryocyte progenitor cells (CFU-Mk). MegaCult™-C medium with cytokines is optimised for the growth of CFU-Mk in human bone marrow, mobilised peripheral blood, and cord blood samples. It is suitable for use with CD34 + enriched cells, mononuclear cells, and cells isolated by other purification methods. The medium contains recombinant human (rh) IL-3, IL-6, and thrombopoietin. Collagen Solution for preparation of collagen gels and for coating cell culture surfaces. Individual kit components may also be purchased separately, for your convenience.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité