MegaCult™-C Complete Kit without Cytokines
Fournisseur: STEMCELL Technologies
04970
STMC04970EA 9056 EUR InStock
STMC04970
MegaCult™-C Complete Kit without Cytokines
Milieux de culture cellulaire
Complete kit including collagen, media, staining kit and chamber slide kit for human CFU-Mk assays.
MegaCult™-C Complete Kit without Cytokines includes all reagents (except cytokines) required for colony-forming unit (CFU) assays of megakaryocyte progenitor cells (CFU-Mk) in a collagen-based culture medium, including the detection of human CFU-Mk colonies by immunocytochemical staining. The addition of appropriate cytokines, as chosen by the user, is required.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité