PneumaCult™-ALI Medium
Fournisseur: STEMCELL Technologies
05001
STMC05001EA 276 EUR InStock
STMC05001
PneumaCult™-ALI Medium
Milieux de culture cellulaire
Serum- and BPE-free medium for human airway epithelial cells cultured at the air-liquid interface.
PneumaCult™-ALI Medium is a serum- and BPE-free medium for the culture of human airway epithelial cells at the air-liquid interface (ALI). Airway epithelial cells cultured in PneumaCult™-ALI Medium undergo extensive mucociliary differentiation to form a pseudostratified epithelium that exhibits morphological and functional characteristics similar to those of the human airway in vivo. PneumaCult™-ALI Medium is also available in a kit that includes 12 mm Transwell® inserts or 6,5 mm Transwell® inserts. Together, PneumaCult™-ALI Medium and PneumaCult™-Ex Plus Medium constitute a fully integrated BPE-free culture system for in vitro human airway modeling that is also compatible with primary human nasal epithelial cells. This robust and defined system is a valuable tool for basic respiratory research, toxicity studies, and drug development.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité