STEMdiff™ Cardiomyocyte Maintenance Kit
Fournisseur: STEMCELL Technologies
05020
STMC05020EA 205 EUR InStock
STMC05020
STEMdiff™ Cardiomyocyte Maintenance Kit
Milieux de culture cellulaire
Medium for long-term maintenance of human PSC-derived cardiomyocytes.
STEMdiff™ Cardiomyocyte Maintenance Kit can be used for long-term maintenance of human pluripotent stem cell (PSC)-derived cardiomyocytes for one month or longer. The functional capacity of hPSC-derived cardiomyocytes is retained for downstream applications and analysis. This kit is compatible with cardiomyocytes generated using STEMdiff™ Ventricular Cardiomyocyte Differentiation Kit or STEMdiff™ Atrial Cardiomyocyte Differentiation Kit.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité