STEMdiff™ Intestinal Organoid Kit
Fournisseur: STEMCELL Technologies
05140
STMC05140EA 1126 EUR InStock
STMC05140
STEMdiff™ Intestinal Organoid Kit
Milieux de culture cellulaire
Serum-free medium system for establishment and maintenance of human intestinal organoids from hPSCs.
STEMdiff™ intestinal organoid kit is a serum-free cell culture medium system that enables robust and efficient generation of human pluripotent stem cell (hPSC)-derived intestinal organoids in a simple, three-stage protocol. Using this kit, hPSCs are directed through induction of definitive endoderm, mid-/hindgut spheroids, and generation of intestinal organoids that can be maintained long-term through passaging or cryopreserved. The organoids exhibit a cellular composition and organization that models the developing intestinal epithelium and associated mesenchyme, making them a convenient model system with direct relevance to the developing intestine. STEMdiff™ intestinal organoid kit is based on the formulation and has been optimised to increase efficiency and reproducibility of organoid formation and expansion across hPSC lines.
Human intestinal organoids can be used as a model system for studying intestinal development and cell biology, intestinal inflammation, intestinal regeneration, microbial interaction, disease modeling, drug discovery, and compound screening. The kit is optimised for differentiation of cells maintained in mTeSR™1. For extended culture and passaging, the kit components required for organoid maintenance can be purchased as STEMdiff™ intestinal organoid growth medium.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité