STEMdiff™ Monocyte Differentiation Supplement (100X)
Fournisseur: STEMCELL Technologies
05324
STMC05324EA 190 EUR InStock
STMC05324
STEMdiff™ Monocyte Differentiation Supplement (100X)
Milieux de culture cellulaire
Supplement for the differention to CD14+ monocytes.
Use STEMdiff™ Monocyte Differentiation Supplement (100X) to support the differention of human pluripotent stem cells (hPSCs) to CD14+ monocytes. Available either individually or as part of the STEMdiff™ Monocyte Kit, the supplement is intended to be combined with StemSpan™ SFEM II to prepare the Monocyte Differentiation Medium. This medium can be used during the STEMdiff™ monocyte differentiation protocol to promote the production of CD14+ monocytes. For detailed information on the STEMdiff™ monocyte differentiation protocol, please explore the STEMdiff™ Monocyte Kit Product Information Sheet.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité