MesenCult™ MSC Basal Medium (Human)
Fournisseur: STEMCELL Technologies
05401
STMC05401EA 79 EUR InStock
STMC05401
MesenCult™ MSC Basal Medium (Human)
Milieux de culture cellulaire
Basal medium for human mesenchymal stem cells.
MesenCult™ MSC basal medium (Human) is a standardised basal medium designed to be supplemented with MesenCult™ mesenchymal stem cell stimulatory supplement (Human) for the in vitro culture of human mesenchymal stem cells (MSCs). MesenCult™ MSC basal medium is a component of MesenCult™ proliferation kit (Human), and is also available separately.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité