MesenCult™ Adipogenic Differentiation Kit (Human)
Fournisseur: STEMCELL Technologies
05412
STMC05412EA 376 EUR InStock
STMC05412
MesenCult™ Adipogenic Differentiation Kit (Human)
Milieux de culture cellulaire
Medium for the differentiation of human MSCs into adipocytes.
MesenCult™ adipogenic differentiation medium (Human) is specifically formulated for the in vitro differentiation of primary human mesenchymal stromal cells, and hPSC-derived mesenchymal progenitor cells (also known as mesenchymal stem cells or MSCs) into adipogenic lineage cells. This kit is suitable for the differentiation of MSCs derived from human bone marrow (BM), adipose tissue, umbilical cord, or pluripotent stem cells (PSCs) that have been previously culture-expanded in serum- and animal component-free medium (e.g. MesenCult™ ACF plus medium), serum-containing medium (e.g. MesenCult™ proliferation kit), or platelet lysate medium (e.g. MesenCult™-hPL medium).
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité