MesenCult™ SF Attachment Substrate
Fournisseur: STEMCELL Technologies
05424
STMC05424EA 555 EUR InStock
STMC05424
MesenCult™ SF Attachment Substrate
Milieux de culture cellulaire
Serum-free attachment substrate for human mesenchymal stem cells - 5 mg.
Support the attachment and growth of human mesenchymal stem and progenitor cell (MSC) cultures in vitro with MesenCult™-SF Attachment Substrate. This serum-free cell culture substrate is optimized to enhance the expansion of human MSCs in vitro and support their enumeration using the colony-forming unit–fibroblast (CFU-F) assay.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further re sale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet ( collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité