MesenCult™ hPL Medium Kit
Fournisseur: STEMCELL Technologies
05439
STMC05439EA 539 EUR InStock
STMC05439
MesenCult™ hPL Medium Kit
Milieux de culture cellulaire
For the culture of human mesenchymal stem cells.
MesenCult™-hPL Medium has been optimised for the culture and expansion of human mesenchymal stromal cells (MSCs; also known as mesencyhymal stem cells) as well as for the detection and enumeration of colony-forming unit fibroblasts (CFU-F). MesenCult™-hPL 10X Supplement is an alternative growth supplement to fetal bovine serum (FBS) that contains purified human platelet lysate (hPL). Complete MesenCult™-hPL Medium (Basal Medium + 10X Supplement) does not require addition of growth factors, lipids, or attachment substrate. It is more defined and provides better performance than FBS-containing media, and is also a more affordable option than defined, serum-free media.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité