MesenCult™ Adipogenic Differentiation Kit (Mouse)
Fournisseur: STEMCELL Technologies
05507
STMC05507EA 396 EUR InStock
STMC05507
MesenCult™ Adipogenic Differentiation Kit (Mouse)
Milieux de culture cellulaire
For the in vitro differentiation of mouse MSCs, ADSCs, and MEFs into adipocytes.
MesenCult™ Adipogenic Differentiation Kit (Mouse) is specifically formulated for the in vitro differentiation of mouse mesenchymal stem and progenitor cells (MSCs), adipose tissue-derived MSCs (ADSCs), and mouse embryonic fibroblasts (MEFs) into cells of the adipogenic lineage. NOTE: MesenCult™ Adipogenic Differentiation Medium must be supplemented with L-glutamine.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité