EpiCult™ B Human Medium Kit
Fournisseur: STEMCELL Technologies
05601
STMC05601EA 332 EUR InStock
STMC05601
EpiCult™ B Human Medium Kit
Milieux de culture cellulaire
For culture and evaluation of human mammary epithelial cells in CFU assays.
EpiCult™-B Human Medium Kit is a serum-free culture medium optimised for the culture of human mammary luminal and myoepithelial cells. It is ideal for the growth and evaluation of bipotent, luminal-restricted and myoepithelial-restricted mammary epithelial progenitor cells in the mammary colony-forming unit (CFU) assay when used in conjunction with an irradiated feeder layer such as NIH 3T3. This kit contains EpiCult™-B Basal Medium (Human) and EpiCult™-B Proliferation Supplement (Human). Addition of Hydrocortisone Stock Solution is required.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité