EpiCult™-B Mouse Medium Kit
Fournisseur: STEMCELL Technologies
05610
STMC05610EA 383 EUR InStock
STMC05610
EpiCult™-B Mouse Medium Kit
Milieux de culture cellulaire
For culture and evaluation of mouse mammary epithelial cells.
EpiCult™-B (Mouse) is a serum-free liquid culture medium optimised for the culture of mouse mammary luminal and myoepithelial cells. It is ideal for the culture and evaluation of mouse mammary epithelial progenitors in the mammary colony-forming unit assay when used in conjunction with an irradiated feeder layer such as NIH 3T3 cells. This medium is also used for enzymatic dissociation of mouse mammary tissue when supplemented with collagenase and hyaluronidase. Addition of human recombinant EGF, human recombinant bFGF, and heparin solution is required for culturing cells.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité