MammoCult™ Human Medium Kit
Fournisseur: STEMCELL Technologies
05620
STMC05620EA 413 EUR InStock
STMC05620
MammoCult™ Human Medium Kit
Milieux de culture cellulaire
For culture of human mammospheres and tumorspheres.
MammoCult™ Medium (Human) is a serum-free, estrogen and progesterone-free culture medium optimised for the culture of mammospheres from normal human primary breast tissues and tumorspheres from human breast cancer cell lines. For preparation of complete MammoCult™ Medium, Hydrocortisone Stock Solution and Heparin Solution are also required.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité