EpiCult™ C Human Medium Kit
Fournisseur: STEMCELL Technologies
05630
STMC05630EA 296 EUR InStock
STMC05630
EpiCult™ C Human Medium Kit
Milieux de culture cellulaire
For culture of human mammary epithelial cells.
EpiCult™-C Medium (Human) is a serum-free culture medium optimised for the short-term culture of human mammary luminal and myoepithelial cells. This medium is also ideal for the enzymatic dissociation of human mammary tissue when supplemented with Collagenase/Hyaluronidase. This kit contains EpiCult™-C Basal Medium (Human) and EpiCult™-C Proliferation Supplement (Human). Addition of Hydrocortisone Stock Solution is required.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité