NeuroCult™ Differentiation Kit (Mouse and Rat)
Fournisseur: STEMCELL Technologies
05704
STMC05704EA 354 EUR InStock
STMC05704
NeuroCult™ Differentiation Kit (Mouse and Rat)
Milieux de culture cellulaire
Medium kit for differentiation of mouse and rat neural stem and progenitor cells.
NeuroCult™ Differentiation Kit (Mouse and Rat) is a standardised medium and supplement kit for the differentiation of mouse and rat neural stem and progenitor cells into neurons, astrocytes, and oligodendrocytes.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité