NeuroCult™ SM1 Neuronal Supplement
Fournisseur: STEMCELL Technologies
05711
STMC05711EA 93 EUR InStock
STMC05711
NeuroCult™ SM1 Neuronal Supplement
Milieux de culture cellulaire
Supplement (50X) for the serum-free culture of neurons.
Avoid culture failure with a Brewer’s B27-based supplement that provides a consistent neuronal culture experience for CNS-derived or human pluripotent stem cell (hPSC)-derived cells. NeuroCult™ SM1 (STEMCELL Modified-1) Neuronal Supplement is based on the published formulation (Brewer et al. J Neurosci Res., 1993) and standardized to more reproducibly support survival and maturation of functional primary and human pluripotent stem cell (hPSC)-derived neurons. This serum-free supplement can be used with basal media and a variety of different induction factors or cytokines to support differentiation along ectoderm, mesoderm and endoderm lineages. NeuroCult™ SM1 may also be used as a serum-replacement supplement for various customizable applications, such as neurotoxicity assays and calcium imaging. For your convenience, NeuroCult™ SM1 is included as a component of multiple BrainPhys™ Neuronal Medium culture kits for primary and hPSC-derived neurons. For further details, see the performance data with BrainPhys™.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité