NeuroCult™ Neuronal Plating Medium
Fournisseur: STEMCELL Technologies
05713
STMC05713EA 62 EUR InStock
STMC05713
NeuroCult™ Neuronal Plating Medium
Milieux de culture cellulaire
Culture medium for plating dissociated primary tissue-derived neurons for improved survival.
NeuroCult™ neuronal plating medium is a serum-free neuronal basal medium. It is designed for use with BrainPhys™ neuronal medium for the plating and culture of primary tissue-derived neurons.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité