NeuroCult™-NS A Basal Medium (Human)
Fournisseur: STEMCELL Technologies
05750
STMC05750EA 100 EUR InStock
STMC05750
NeuroCult™-NS A Basal Medium (Human)
Milieux de culture cellulaire
Basal medium for expansion of human neural stem and progenitor cells.
NeuroCult™ NS-A Basal Medium (Human) is a standardised basal medium for the expansion of human neural stem cells and brain tumor stem cells, in the neurosphere or adherent monolayer culture system. This basal medium is a component of the NeuroCult™ NS-A Proliferation Kit and the NeuroCult™ NS-A Differentiation Kit).
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité