NeuroCult™-XF Proliferation Medium
Fournisseur: STEMCELL Technologies
05761
STMC05761EA 382 EUR InStock
STMC05761
NeuroCult™-XF Proliferation Medium
Milieux de culture cellulaire
Xeno-free medium for expansion of human neural stem and progenitor cells.
NeuroCult™-XF Proliferation Medium is a standardised, xeno-free, serum-free medium for the culture of human neural stem and progenitor cells from normal tissues or tumor samples, in the neurosphere or adherent monolayer system. When supplemented with appropriate cytokines, NeuroCult™-XF Proliferation Medium is optimised to maintain human neural stem cells in culture for extended periods of time without the loss of their self-renewal, proliferation or differentiation potential.
NOTE: Addition of Carrier-Free rh EGF and rh bFGF is required. Heparin can also be added, however, the heparin solution contains non-human animal-derived components and can be omitted if a complete xeno-free system is desired.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité