BrainPhys™ Neuronal Medium and SM1 Kit
05792
STMC05792
BrainPhys™ Neuronal Medium and SM1 Kit
Milieux de culture cellulaire
Kit including BrainPhys™ Neuronal Medium and SM1 Neuronal Supplement for culture of primary and ES/iPS cell-derived neurons.
Culture primary or human pluripotent stem cell (hPSC)-derived neurons long term in a medium optimised to promote, rather than inhibit neuronal activity and maturity. For your convenience, BrainPhys™ Neuronal Medium and SM1 Kit includes both serum-free BrainPhys™ Neuronal Medium (basal medium) and NeuroCult™ SM1 Neuronal Supplement, a Brewer's B27-based supplement that ensures cell health and encourages neurite outgrowth and branching in short- and long-term serum-free cultures.
Based on the formulation, BrainPhys™ Neuronal Medium mimics the extracellular environment of the central nervous system (CNS) to yield a higher proportion of synaptically active neurons. Use BrainPhys™ Neuronal Medium and SM1 Kit to culture primary CNS-derived neurons or embryonic stem (ES) and induced pluripotent stem (iPS) cell-derived neurons. To avoid shocking cells with media changes, also use this culture system when performing functional assays, such as microelectrode array-based recordings or live-fluorescent imaging.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité