BrainPhys™ hPSC Neuron Kit
Fournisseur: STEMCELL Technologies
05795
STMC05795EA 742 EUR InStock
STMC05795
BrainPhys™ hPSC Neuron Kit
Milieux de culture cellulaire
Kit including BrainPhys™ Neuronal Medium, SM1 Neuronal Supplement, N2 Supplement-A, BDNF and GDNF for culture of ES/iPS cell-derived neurons.
Culture, differentiate, and mature neurons derived from human embryonic stem (ES) or induced pluripotent stem (iPS) cells by using a complete medium optimised to promote, rather than inhibit neuronal activity. For your convenience, BrainPhys™ hPSC Neuron Kit includes serum-free BrainPhys™ Neuronal Medium (basal medium), supplements, and growth factors to enable generation and maturation of different neuronal subtypes from human ES/iPS cell-derived neural progenitor cells. Based on the formulation, BrainPhys™ Neuronal Medium mimics the extracellular environment of the central nervous system (CNS) to yield a higher proportion of synaptically active neurons.
NeuroCult™ SM1 Neuronal Supplement ensures cell health and encourages neurite outgrowth and branching in short- and long-term serum-free cultures, and N2 Supplement-A supports the in vitro differentiation of ES/iPS-derived cells to neuronal subtypes. Included BDNF and GDNF growth factors support lineage-specific differentiation. To avoid shocking cells with media changes, also use BrainPhys™ medium when performing functional assays, such as microelectrode array-based recordings or live-fluorescent imaging. View our additional resources to learn more about the BrainPhys™ system.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité