BrainPhys™ Imaging Optimised Medium
Fournisseur: STEMCELL Technologies
05796
STMC05796EA 154 EUR InStock
STMC05796
BrainPhys™ Imaging Optimised Medium
Milieux de culture cellulaire
Serum-free neurophysiological basal medium for improved neuronal live imaging and function.
BrainPhys™ Imaging optimised Medium is a serum-free neuronal basal medium that is optimised for imaging applications. Applications of BrainPhys™ Imaging optimised Medium include live fluorescent imaging (calcium imaging and optogenetics) and neuronal culture. In addition to the removal of phenol red, the formulation has been modified to reduce background fluorescence and increase stability upon repeated exposure to light.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité