STEMdiff™ Neural Progenitor Medium
05833
STMC05833
STEMdiff™ Neural Progenitor Medium
Milieux de culture cellulaire
Medium for maintenance and expansion of neural progenitor cells derived from human ES and iPS cells.
STEMdiff™ neural progenitor medium is a defined and serum-free medium for the expansion of neural progenitor cells (NPCs) derived from human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells using STEMdiff™ neural induction medium. NPCs cultured in this medium can be expanded 3 to 5 fold per passage, and cultured for at least 10 passages, with minimal spontaneous neuronal differentiation.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité